Läs mer
A66, MedChemExpress
Produktkod. 16475598
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16475598 10 mg
16461767 50 mg
16448297 5 mg
16406146 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16475598 Leverantör Medchem Express Leverantörsnummer HY13261
MedChemExpress A66 is a highly specific and selective p110α inhibitor with an IC50 of 32 nM.
A66 is a highly specific and selective p110α inhibitor with an IC50 of 32 nM.
Specifikationer
Kemiskt namn eller material A66
CAS 1166227-08-2
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C17H23N5O2S2
Kvantitet 10 mg
Löslighetsinformation DMSO : 50 mg/mL (127.06 mM; Need ultrasonic)
LEDER CC(C)(C1=NC(C2=C(N=C(S2)NC(N3[C@@H](CCC3)C(N)=O)=O)C)=CS1)C
Molekylvikt (g/mol) 393.53
Formel vikt 393.53
Procent renhet 98.44%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease