Läs mer
Tocris Bioscience™ AA 147
Beskrivning
AA 147 is an endoplasmic reticulum (ER) proteostasis regulator. Activates the ATF6 transcriptional program independently of ER stress and/or global UPR activation. Selectively reduces secretion of amyloidogenic transthyretin (TTR) from HepG2 cells. Reduces amyloidogenic immunoglobulin light chain (ALLC) secretion from ALMC-2 cells (EC50 = 1.1 μM).
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-(2-Hydroxy-5-methylphenyl)benzenepropanamide |
| CAS | 393121-74-9 |
| Kvantitet | 25 mg |
| Molekylformel | C16H17NO2 |
| Formel vikt | 255.31 |
| Renhet | 98% |