Läs mer
Abamectin A2a (>80%), TRC
Produktkod. 30180209
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180209 1 mg
30180131 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180209 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1070151MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 65195-53-1
Molekylformel C49H76O15
Molekylvikt (g/mol) 905.12
Synonym Avermectin A2a
LEDER CC[C@H](C)[C@H]1O[C@@]2(C[C@@H]3CC(C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@@H](O)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C\6/CO[C@@H]7[C@H](OC)C(=C[C@@H](C(=O)O3)[C@]67O)C)O2)C[C@H](O)[C@@H]1C
Specifikationer
Kemiskt namn eller material Avermectin A2a