Learn More
Abcam Donkey F(ab')2 Anti-Mouse IgG H&L (Alexa Fluor™ 647), 500 ug
Product Code. 32111007 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
Product Code. Quantity Unit Size
32111007 500 μg 500µg
1 options
Product Code. 32111007 Supplier Abcam Supplier No. ab181292500ug
 Restricted Item
Beräknad leverans: 02-12-2026
Donkey Polyclonal secondary antibody, suitable for ELISA, IHC-P, Flow Cyt, ICC/IF, with 4 citations.
  • Host Species: Donkey
  • Clonality: Polyclonal
  • Applications: ELISA, IHC-P, Flow Cyt, ICC/IF
  • Species Reactivity: Mouse
  • Storage Buffer: Preservative: 0.02% Sodium azide, Constituents: PBS, 23% Glycerol (glycerin, glycerine), 1% BSA
  • Form: Liquid
  • Purification: Affinity purification Immunogen
Specifications
Applications ELISA, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation Preservative: 0.02% Sodium azide, Constituents: PBS, 23% Glycerol (glycerin, glycerine), 1% BSA
Gene Symbols Ighg1
Host Species Donkey
Purification Method Affinity purification Immunogen
Quantity 500 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Polyclonal Secondary Antibody
Form Liquid
Shipping Condition Blue Ice
Reactivity abrv Ms
Validated Application abrv ELISA, IHC-P, Flow Cyt, ICC/IF