Learn More
Abcam Human Kallikrein 14 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Product Code. 32113273 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
96 tests
Product Code. Quantity Unit Size
32113273 10 x 96 Tests 96 tests
1 options
Product Code. 32113273 Supplier Abcam Supplier No. ab25359010x96Tests
 Restricted Item
Beräknad leverans: 26-11-2026
Antibody, suitable for sELISA.
  • Applications: sELISA
  • Species Reactivity: Human
Specifications
Applications Sandwich ELISA
Conjugate Unconjugated
Gene Symbols KLK14
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Human.
Target Species Human
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Hu
Validated Application abrv sELISA