Learn More
Abcam Human TNF alpha Matched Antibody Pair Kit, 5 x 96 Tests
Product Code. 32120014 Shop All Abcam Products
Change view
Click to view available options
Quantity:
5 x 96 Tests
Unit Size:
96 tests
Product Code. Quantity Unit Size
32120014 5 x 96 Tests 96 tests
1 options
Product Code. 32120014 Supplier Abcam Supplier No. ab2134675x96Tests
 Restricted Item
Beräknad leverans: 26-11-2026
Antibody, suitable for ELISA, IA, with 2 citations.
  • Applications: ELISA, IA
  • Species Reactivity: Human
Specifications
Applications ELISA
Conjugate Unconjugated
Gene Symbols TNF
Quantity 5 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Human.
Target Species Human
Product Type Secondary Antibody
Detection Method Colorimetric
Shipping Condition Blue Ice
Reactivity abrv Hu
Validated Application abrv ELISA, IA