Learn More
Abcam mCherry Antibody Pair - BSA and Azide free, 10 x 96 Tests
Product Code. 32112445 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
96 tests
Product Code. Quantity Unit Size
32112445 10 x 96 Tests 96 tests
1 options
Product Code. 32112445 Supplier Abcam Supplier No. ab24399910x96Tests
 Restricted Item
Beräknad leverans: 26-11-2026
Antibody, suitable for sELISA.
  • Applications: sELISA
  • Species Reactivity: Bacteria
Specifications
Antigen mCherry
Applications Sandwich ELISA
Conjugate Unconjugated
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Bacteria.
Target Species Bacteria
Product Type Secondary Antibody
Shipping Condition Blue Ice
Validated Application abrv sELISA