missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ ABCB7 Polyclonal Antibody
GREENER_CHOICE

Produktkod. 16627924
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16627924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16627924 Leverantör Invitrogen™ Leverantörsnummer PA5101445

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ABCB7.

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. This gene encodes a half-transporter involved in the transport of heme from the mitochondria to the cytosol. With iron/sulfur cluster precursors as its substrates, this protein may play a role in metal homeostasis. Mutations in this gene have been implicated in X-linked sideroblastic anemia with ataxia.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen ABCB7
Användningsområden Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gen ABCB7
Genaccessionsnr. O75027, Q61102
Gene Alias AA517758; ABC transporter 7 protein; ABC7; ABCB7; ASAT; Atm1p; ATP binding cassette subfamily B member 7; ATP-binding cassette 7; ATP-binding cassette sub-family B member 7; ATP-binding cassette sub-family B member 7, mitochondrial; ATP-binding cassette sub-family B member 7, mitochondrial-like protein; ATP-binding cassette transporter 7; ATP-binding cassette, subfamily B (MDR/TAP), member 7; ATP-binding cassette, sub-family B (MDR/TAP), member 7; ATP-binding cassette, sub-family B, member 7; ATP-binding cassette, sub-family B, member 7, mitochondrial; AU019072; EST140535; fc20c02; wu:fc20c02
Gensymboler ABCB7
Värd art Rabbit
Immunogen A synthesized peptide derived from human ABCB7(Accession O75027), corresponding to amino acid residues F77-Y127.
Reningsmetod Affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 11306, 22
Målarter Human, Mouse
Innehåll och lagring -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.