missing translation for 'onlineSavingsMsg'
Learn More

ABCF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13950-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16832394

  • 1656.73 SEK / 20µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a component of the spliceosome complex and is one of several retinitis pigmentosa-causing genes. When the gene product is added to the spliceosome complex, activation occurs.
Specifications

Specifications

ABCF1
Polyclonal
Unconjugated
ABCF1
ABC27, ABC50, ABCF1, ATP binding cassette 50
Rabbit
Antigen Affinity Chromatography
RUO
224742, 23, 85493
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6MG08, Q6P542, Q8NE71
Abcf1
ABCF1 Fusion Protein Ag5061
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers