missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
ABCG1 Antibody, PE/Cy7, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-132PECY7
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
ABCG1 Polyclonal specifically detects ABCG1 in Human, Mouse, Rat, Chinese Hamster, Hamster, Monkey, Rabbit samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifikationer
| ABCG1 | |
| Polyclonal | |
| Flow Cytometry, Flow (Intracellular) | |
| ABC transporter 8, ABC8ATP-binding cassette sub-family G member 1, ATP-binding cassette transporter 8, ATP-binding cassette, sub-family G (WHITE), member 1, homolog of Drosophila white, MGC34313, White protein homolog, white protein homolog (ATP-binding cassette transporter 8), 10ATP-binding cassette transporter member 1 of subfamily G, WHITE1, WHT1 | |
| Rabbit | |
| Affinity Purified | |
| ABC Transporters, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 9619.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| ABCG1 | |
| A synthetic peptide made to an internal region of human ABCG1 (between residues 300-400). [UniProt No. P45844] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Chinese Hamster, Hamster, Monkey, Rabbit | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering