Learn More
CleanSpace Technology™ Combined Filter
Shop All CleanSpace Technology Products
Click to view available options
Model:
A1 P3 TM3 SLR
A2 P3 P SL R
TM3 ABE1 P SL R
Unit Size:
Each
Description
- Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
- Materials: Fibreglass particulate media, activated carbon and plastic casing, silicone seal
- Easily fitted and removed from the power unit
Specifications
Specifications
Certifications/Compliance | EN 12942 |
Product Type | Filter |
For Use With (Equipment) | CleanSpace PAPR Power Units |
Model | TM3 ABE1 P SL R |
Product Content Correction
Product Title
CleanSpace Technology™ Combined Filter >
Spot an opportunity for improvement?Share a Content Correction