missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ ABHD12 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA518982
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody is predicted to react with human based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene encodes an enzyme that catalyzes the hydrolysis of 2-arachidonoyl glycerol, the main endocannabinoid lipid transmitter that acts on cannabinoid receptors, CB1 and CB2. The endocannabinoid system is involved in a wide range of physiological processes, including neurotransmission, mood, appetite, pain appreciation, addiction behavior, and inflammation. Mutations in this gene are associated with the neurodegenerative disease, PHARC, resulting from an inborn error of endocannabinoid metabolism. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Specifikationer
| ABHD12 | |
| Polyclonal | |
| Unconjugated | |
| ABHD12 | |
| 1500011G07Rik; 2-arachidonoylglycerol hydrolase; 2-arachidonoylglycerol hydrolase ABHD12; 6330583M11Rik; Abhd12; ABHD12A; abhydrolase domain containing 12; abhydrolase domain-containing protein 12; AI431047; AW547313; BEM46L2; C20orf22; dJ965G21.2; DKFZP434P106; hABHD12; Lysophosphatidylserine lipase ABHD12; Monoacylglycerol lipase ABHD12; Oxidized phosphatidylserine lipase ABHD12; PHARC | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 26090, 499913, 76192 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q6AYT7, Q8N2K0, Q99LR1 | |
| ABHD12 | |
| Synthetic peptide sequence (REFLGKSEPEHQH) corresponding to the C-terminus amino acids of ABHD12 (aa 386-398). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering