Läs mer
Abiraterone N-Oxide, TRC
Produktkod. 30321593
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30321593 5 mg
1 options
Produktkod. 30321593 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1085405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Abiraterone N-Oxide
CAS 2378463-76-2
Molekylformel C24 H31 N O2
Kvantitet 5 mg
Synonym Androsta-5,16-dien-3-ol, 17-(1-oxido-3-pyridinyl)-, (3β)- (ACI), Abiraterone N-oxide, Androsta-5,16-dien-3-ol, 17-(1-oxido-3-pyridinyl)-, (3β)- (ACI), (3β)-17-(1-Oxido-3-pyridinyl)androsta-5,16-dien-3-ol (ACI)
InChI formel InChI=1S/C24H31NO2/c1-23-11-9-18(26)14-17(23)5-6-19-21-8-7-20(16-4-3-13-25(27)15-16)24(21,2)12-10-22(19)23/h3-5,7,13,15,18-19,21-22,26H,6,8-12,14H2,1-2H3/t18-,19-,21-,22-,23-,24+/m0/s1
LEDER C[C@]12CC[C@H]3[C@@H](CC=C4C[C@@H](O)CC[C@]34C)[C@@H]1CC=C2c5ccc[n+]([O-])c5
IUPAC-namn (3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-(1-oxidopyridin-1-ium-3-yl)-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-ol
Molekylvikt (g/mol) 365.5084
Formel vikt 365.2355