Läs mer
Abiraterone, TRC
Produktkod. 30009259
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009259 100 mg
18030740 5 mg
18003230 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009259 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA108490100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 154229-19-3
Molekylformel C24 H31 N O
Molekylvikt (g/mol) 349.51
Synonym (3β)-17-(3-Pyridinyl)androsta-5,16-dien-3-ol, 17-(3-Pyridinyl)-androsta-5,16-dien-3β-ol, Abiraterone, CB 7598
IUPAC-namn (3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-pyridin-3-yl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-ol
LEDER C[C@]12CC[C@H]3[C@@H](CC=C4C[C@@H](O)CC[C@]34C)[C@@H]1CC=C2c5cccnc5
Specifikationer
Kemiskt namn eller material Abiraterone
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C24H31NO/c1-23-11-9-18(26)14-17(23)5-6-19-21-8-7-20(16-4-3-13-25-15-16)24(21,2)12-10-22(19)23/h3-5,7,13,15,18-19,21-22,26H,6,8-12,14H2,1-2H3/t18-,19-,21-,22-,23-,24+/m0/s1
Formel vikt 349.2406
Procent renhet >95%