missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ A2M (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human A2M in Plasma and Serum
Brand: Abnova™ KA0486.1Kit
Additional Details : Weight : 0.02000kg
Specifications
1.25 to 40μg/mL | |
Human | |
1μg/mL | |
Quantification | |
A2M (Human) ELISA Kit | |
1 Kit | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Biotin | |
A2M (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human A2M in cell culture supernatant. | |
Colorimetric | |
96-well Polystyrene microplate coated with Polyclonal Antibody, precut, pressure sensitive sealing tapes, 20μg Human a-2-macroglobulin in a Buffered protein base, 1 vial Biotinylated a-2-Macroglobulin, lyophilized, 30mL 10-fold EIA Dilue | |
A2m (Human) ELISA kit | |
Plasma, Serum | |
50μL |