missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ A2M (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human A2M in Plasma and Serum
POA
Specifications
Calibration Range | 1.25 to 40μg/mL |
---|---|
Detection Limit | 1μg/mL |
Detection Method | Colorimetric |
Conjugate | Biotin |
Cross Reactivity | Human |
Includes:
- Polystyrene microplate coated with polyclonal antibody against α - 2 - Macroglobulin, 96 (8 x 12) wells
- Pre - cut, pressure - sensitive sealing tapes, which can be cut to fit the format of the individual assay, 3 in no.
- Human α - 2 - Macroglobulin in a buffered protein base (lyophilized), 20μg
- Biotinylated α-2-Macroglobulin, lyophilized, 1 vial
- 10-fold EIA diluent concentrated buffered protein base, 30mL
- 20-fold concentrated wash buffered surfactant, 30mL
- 100-fold Streptavidin-Peroxidase conjugate (SP conjugate), 80μL
- Ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine, 8mL
- 0.5N hydrochloric acid to stop the chromogen substrate reaction, 12mL
Specifications
1.25 to 40μg/mL | |
Colorimetric | |
Human | |
Quantification | |
1 Kit | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
1μg/mL | |
Biotin | |
A2M (Human) ELISA Kit | |
96-well Polystyrene microplate coated with Polyclonal Antibody, precut, pressure sensitive sealing tapes, 20μg Human a-2-macroglobulin in a Buffered protein base, 1 vial Biotinylated a-2-Macroglobulin, lyophilized, 30mL 10-fold EIA Dilue | |
Plasma, Serum |