missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ADIPOQ (Human) ELISA Kit
For detection of ADIPOQ in Human Urine and Murine, Adiponectin in Plasma, Serum and cell culture supernatants
Brand: Abnova™ KA0978.1Kit
Additional Details : Weight : 0.02000kg
Specifications
0.78 to 50ng/mL | |
Human | |
0.7ng/mL | |
Quantification | |
ADIPOQ (Human) ELISA Kit | |
RUO | |
Store components of the kit at 4°C or -20°C as described in the protocol. | |
50μL |
Biotin | |
ADIPOQ (Human) ELISA Kit is designed for detection of ADIPOQ in human urine and murine, adiponectin in plasma, serum and cell culture supernatants. | |
Colorimetric | |
96-well Antibody coated microwell, 800ng Human adiponectin standard, 1 vial Biotinylated adiponectin Antibody (100X), 30mL mix diluent concentrate (10X), 30mL x 2 Wash Buffer 20X, 80 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL chromogen Substrate, 12mL stop solution | |
96 Assays | |
Cell Culture Supernatant, Milk, Plasma, Saliva, Serum, Urine | |
96 Assays |