missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Adipoq (Mouse) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Adipoq in Urine, Plasma, Serum and cell culture supernatants
Brand: Abnova™ KA0018.1Kit
Additional Details : Weight : 0.02000kg
Specifications
0.25 to 8ng/mL | |
Mouse | |
0.079ng/mL | |
Quantification | |
Adipoq (Mouse) ELISA Kit | |
1 Kit | |
Cell Culture Supernatant, Serum | |
96 Assays |
HRP | |
Adipoq (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Adiponectin/Acrp30 protein in serum and tissue culture medium. | |
Colorimetric | |
96 wells Antigen coated microtiter strips, Conjugate solution (13mL), Master standards (2vials), Quality control HIGH (2vials), Quality control LOW (2vials), Dilution buffer (22mL), Wash solution conc. 10X (100mL), Substrate solution (13mL), Stop solution (13mL) | |
ADIPOQ (Mouse) ELISA kit | |
RUO | |
Store the kit at 4°C. | |
100μL |