missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Adipoq (Mouse) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Adipoq in Urine, Plasma, Serum and cell culture supernatants
POA
Specifications
Detection Method | Colorimetric |
---|---|
Cross Reactivity | Mouse |
Product Type | Adipoq (Mouse) ELISA Kit |
Quantity | 1 Kit |
Regulatory Status | For research use only (RUO) |
Specifications
Colorimetric | |
Adipoq (Mouse) ELISA Kit | |
For research use only (RUO) |
Mouse | |
1 Kit |