missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Albumin (Pig) ELISA Kit
Competitive Enzyme Immunoassay for the quantitative measurement of pig ALB in Plasma and Serum
POA
Specifications
Detection Method | Colorimetric |
---|---|
Cross Reactivity | Pig |
Product Type | Albumin (Pig) ELISA kit |
Quantity | 1 Kit |
Regulatory Status | For research use only (RUO) |
Specifications
Colorimetric | |
Albumin (Pig) ELISA kit | |
For research use only (RUO) |
Pig | |
1 Kit | |
Store components of the kit at 4°C or -20°C as described in the protocol. |