missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ANGPT2 (Human) ELISA Kit
In-Vitro Enzyme-Linked Iimmunosorbent assay for the quantitative measurement of Human ANGPT2
POA
Specifications
Detection Method | Colorimetric |
---|---|
Cross Reactivity | Human |
Quantity | 96 Assays |
Regulatory Status | For research use only (RUO) |
Content And Storage | Store the kit at 4°C. Standard should be stored at -80°C after reconstitution. Avoid repeated freezing and thawing. |
Specifications
Colorimetric | |
96 Assays | |
Store the kit at 4°C. Standard should be stored at -80°C after reconstitution. Avoid repeated freezing and thawing. |
Human | |
For research use only (RUO) |