missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ANGPT2 (Human) ELISA Kit
In-Vitro Enzyme-Linked Iimmunosorbent assay for the quantitative measurement of Human ANGPT2
Brand: Abnova™ KA1867.1Kit
Additional Details : Weight : 0.02000kg
Specifications
Biotin | |
ANGPT2 (Human) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human ANGPT2. | |
Colorimetric | |
96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP-streptavidian concentrate, 12mL TMB one-step Substrate reagent, 8mL stop solut | |
96 Assays | |
Cell Culture Supernatant, Plasma, Serum, Urine | |
96 assays |
Human | |
10pg/mL | |
Quantification | |
ANGPT2 (Human) ELISA Kit | |
For research use only (RUO) | |
Store the kit at 4°C. Standard should be stored at -80°C after reconstitution. Avoid repeated freezing and thawing. | |
100μL |