missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ B2M (Human) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Human B2M in Plasma, Serum, Urine and cell culture supernatants

POA

Specifications

Detection Method Colorimetric
Product Type B2M (Human) ELISA kit
View More Specs

Includes:
  • β2-Microglobulin Microplate: A polystyrene microplate coated with a polyclonal antibody against human β2-Microglobulin, 96 (8 x 12) wells
  • Pre-cut, pressure-sensitive sealing tapes, which can be cut to fit the format of the individual assay, 3
  • Human β2-Microglobulin Standard: Human β2-Microglobulin in a buffered proteinbase (lyophilized), 200ng
  • Biotinylated Human β2-Microglobulin Antibody: A 100-fold biotinylated polyclonal antibody against β2-Microglobulin, 80μL
  • A 10-fold MIX Diluent concentrated buffered protein base, 30mL
  • A 20-fold concentrated wash buffered surfactant, 30mL x 2
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate, 80μL
  • Ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine, 8mL
  • Stop Solution: 0.5N hydrochloric acid to stop the chromogen substrate reaction, 12mL

Description

Description

Specifications

Specifications

Colorimetric
B2M (Human) ELISA kit
SDS