missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ B2M (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human B2M in Plasma, Serum, Urine and cell culture supernatants
Brand: Abnova™ KA1045.1Kit
Additional Details : Weight : 0.02000kg
Specifications
0.049 to 50ng/mL | |
Human | |
0.04ng/mL | |
Quantification | |
96-well Antibody coated microwell, 200ng Human B2M standard, 100X Biotinylated B2M Antibody 1 vial, 30mL 10X MIX diluent concentrate, 30mL Wash Buffer 20X concentrate 2, Streptavidin peroxidase Conjugate (SP Conjugate) 80, 8mL chromogen Substrate, 12mL stop solution | |
B2M (Human) ELISA kit | |
Cell Culture Supernatant, Cerebrospinal Fluid, Milk, Plasma, Saliva, Serum, Urine | |
50μL |
HRP | |
B2M (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human B2M. | |
Colorimetric | |
Liquid | |
B2M (Human) ELISA Kit | |
1 Kit | |
Store components of the kit at 4°C or -20°C as described in the protocol. |