Learn More
Abnova™ Caspase Substrate Assay Kit Plus (Fluorometric)
Brand: Abnova™ KA3697.1Kit
Additional Details : Weight : 0.02000kg
Specifications
Caspase Substrate Assay Kit (Fluorometric) is used for assaying activities of members of caspase 1/2/3/5/6/8/9. | |
Functional Study | |
Cell Culture |
Fluorometric | |
1 Kit | |
Store the kit at -20°C. |