missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CCL7 (Human) ELISA Kit
Brand: Abnova™ KA0519.1Kit
Additional Details : Weight : 0.02000kg
Includes: MCP-3 Microtiter plate, 96 wells
MCP-3 Conjugate, 12mL
MCP-3 Standard, 2 vials
Calibrator diluent I, 25mL
Calibrator diluent II, 25mL
Wash Buffer (20x), 60mL
Substrate A, 10mL
Substrate B, 10mL
Stop solution, 14mL
Description
This MCP-3 enzyme linked immunosorbent assay (ELISA) applies a technique called a quantitative sandwich immunoassaySpecifications
30 to 1600pg/mL | |
Human | |
30pg/mL | |
Quantification | |
Antibody coated microwells (96 wells), MCP-3 CONJUGATE (15mL), MCP-3 STANDARD (6.4ng/vial × 2), Calibrator diluent I (22mL), Calibrator diluent II (22mL), Sample diluent (12mL), Wash buffer 20X (60mL), Substrate A and B (11mL each), Stop solution (14mL) | |
ELISA Kit | |
RUO | |
100μL | |
Store the kit at 4°C. | |
100μL |
HRP | |
CCL7 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human CCL7. | |
Colorimetric | |
96 wells | |
CCL7 (Human) ELISA Kit | |
1 Kit | |
Cell Culture Supernatant, Plasma, Serum | |
With Calibrator Diluent I is 30pg/mL, with Calibrator Diluent II is 20pg/mL | |
96 Assays |