Läs mer
Abnova™ CD274/CEN9q (Texas Red/FITC) FISH Probe
Kvantitet:
200 μl
Beskrivning
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifikationer
Specifikationer
| Innehåll och lagring | Store at 4°C in the dark. |
| Beskrivning | Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique. |
| Etikett eller färgämne | FITC, Texas Red |
| För användning med (applikation) | DNA Hybridization |
| Plats | 9p24/9q21 |
| Sond | Approx. 280kb/490kb |
| Produkttyp | CD274(PD-L1)/CEN9q FISH Probe |
| Kvantitet | 200 μl |
| Källa | Genomic DNA |
| Art | Human |