Learn More
Abnova™ CEN8q (R6G) FISH Probe
Labeled FISH probes as chromosomal markers using Fluorescent In Situ Hybridization Technique.
Brand:  Abnova™ FC0068.20uL
Additional Details : Weight : 0.02000kg
Product Code. 16052645
  • 8282.00 SEK / 20µL
Estimated Shipment
04-10-2019
Add to basket
Includes: FISH Hybridization Buffer (80μL)
Description
Description
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Specifications
R6G
FISH Hybridization Buffer (80 uL)
CEN8q
20 uL
Fluorescent In Situ Hybridization (Cell)
8q11.21
CEN8q (R6G) FISH Probe
Store at 4°C in the dark.