Learn More
Abnova™ COL18A1 (Human) ELISA Kit
In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human COL18A1
Brand:  Abnova™ KA3409.1Kit
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16057985
  • 5943.00 SEK / 1 set
Estimated Shipment
03-02-2021
Add to basket
Specifications
156 to 10000pg/mL
Human
<10pg/mL
Quantification
COL18A1 (Human) ELISA Kit
1 Kit
Cell Culture Supernate, Human Milk, Plasma (EDTA, Heparin), Serum
96 Assays
Biotin
COL18A1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human COL18A1.
Colorimetric
96-well plate pre-coated with anti-Human Endostatin antibody, Lyophilized Endostatin standards (10ng x 2), Sample diluent buffer (30mL), Biotin conjugated anti-Human Endostatin antibody (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (130uL), Antibody diluent buffer (12mL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL)
COL18A1 (Human) ELISA kit
RUO
Store the kit at-20°C.
100μL