missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ COL18A1 (Human) ELISA Kit
In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human COL18A1
POA
Specifications
Detection Method | Colorimetric |
---|---|
Conjugate | Biotin |
Cross Reactivity | Human |
Product Type | COL18A1 (Human) ELISA Kit |
Quantity | 96 Assays |
Specifications
Colorimetric | |
Human | |
96 Assays | |
100 μL |
Biotin | |
COL18A1 (Human) ELISA Kit | |
For research use only (RUO) |