missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Cortisol (Sheep) ELISA Kit
Immunoassay for the quantitative determination of cortisol in sheep serum, plasma, urine
Brand: Abnova™ KA2317.1Kit
Additional Details : Weight : 0.02000kg
Specifications
0 to 200ng/mL | |
Sheep | |
1.0ng/mL | |
Quantification | |
Cortisol (Sheep) ELISA Kit | |
1 Kit | |
Plasma, Serum, Urine | |
96 Assays |
HRP | |
Cortisol (Sheep) ELISA Kit is an immunoassay for the quantitative determination of cortisol in sheep serum, plasma, urine. | |
Colorimetric | |
96 wells-Microtiter wells coated with cortisol specific antibody, HRPEnzyme Conjugate (12mL), Cortisol Standards 1 sets (0.5mL/vial), TMB Color reagent (12mL), Stop solution (6mL), Wash buffer 20X (25mL), Standard diluent (20mL) | |
Cortisol (Sheep) ELISA kit | |
RUO | |
Store the kit at 4°C. | |
50μL |