Learn More
Abnova™ CXCL1 (Human) ELISA Kit
In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human CXCL1
Brand:  Abnova™ KA3337.1Kit
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16057925
  • 5943.00 SEK / 1 set
Estimated Shipment
11-02-2021
Add to basket
Specifications
15.6 to 1000pg/mL
CXCL1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human CXCL1.
Quantification
CXCL1 (Human) ELISA kit
Cell Culture Supernate, Plasma (Citrate, EDTA, Heparin), Serum
Store the kit at-20°C.
Human
Colorimetric
CXCL1 (Human) ELISA Kit
1 Kit
100μL