missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Cxcl2 (Mouse) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Cxcl2
POA
Specifications
Calibration Range | 15.6 to 1000pg/mL |
---|---|
Detection Method | Colorimetric |
Product Type | CXCL2 (Mouse) ELISA kit |
For Use With (Application) | Quantification |
Content And Storage | Store the kit at -20°C |
Includes:
- Lyophilized recombinant mouse MIP-2 standard, 10ng x 2
- Plate precoated with anti-mouse MIP-2 antibody, 96-wells
- Sample diluent buffer, 30mL
- Biotinylated anti-mouse MIP-2 antibody, dilution 1:100, 130μL
- Antibody diluent buffer, 12mL
- Avidin-Biotin-Peroxidase complex (ABC), dilution 1:100, 130μL
- ABC diluent buffer, 12mL
- TMB color delevoping agent, 10mL
- TMB stop solution, 10mL
Specifications
15.6 to 1000pg/mL | |
CXCL2 (Mouse) ELISA kit | |
Store the kit at -20°C |
Colorimetric | |
Quantification |