missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Cxcl2 (Mouse) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Cxcl2
Brand: Abnova™ KA1505.1Kit
Additional Details : Weight : 0.02000kg
Specifications
15.6 to 1000pg/mL | |
Mouse | |
Colorimetric | |
96-well Antibody coated microwell, 10ng x 2 vials lyophilized recombinant Mouse Cxcl2 standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Mouse Cxcl2 Antibody, 12mL Antibody Diluent Buffer, 130μL Avidin-Biotin | |
CXCL2 (Mouse) ELISA kit | |
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate | |
100μL |
HRP | |
Cxcl2 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Cxcl2. | |
Quantification | |
Cxcl2 (Mouse) ELISA Kit | |
1 Kit | |
Store the kit at-20°C. |