missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ENO2 (Human) ELISA Kit
Enzyme Immunoassay for the quantitative measurement of ENO2 in Serum
POA
Specifications
Detection Method | Colorimetric |
---|---|
Product Type | ENO2 (Human) ELISA kit |
Quantity | 1 Kit |
Regulatory Status | For research use only (RUO) |
Content And Storage | Store the kit at 4°C |
Specifications
Colorimetric | |
1 Kit | |
Store the kit at 4°C |
ENO2 (Human) ELISA kit | |
For research use only (RUO) |