missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Epstein Barr Virus VCA IgG ELISA Kit
Solid phase Enzyme Immunoassay for the quantitative measurement of IgG class autoantibodies against Epstein Barr Virus VCA
POA
Specifications
Detection Method | Colorimetric |
---|---|
Conjugate | HRP |
Product Type | Epstein Barr Virus VCA IgG ELISA Kit |
Quantity | 1 Kit |
Specifications
Colorimetric | |
HRP | |
Epstein Barr Virus VCA IgG ELISA Kit | |
1 Kit |