missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Fibrinogen (Human) ELISA Kit
POA
Specifications
Detection Method | Colorimetric |
---|---|
Conjugate | Biotin |
Cross Reactivity | Human |
Product Type | ELISA Kit |
Regulatory Status | For research use only (RUO) |
Description
Enzyme immunoassay for the quantitative measurement of human fibrinogen.Specifications
Colorimetric | |
Human | |
For research use only (RUO) |
Biotin | |
ELISA Kit |