missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Fibrinogen (Human) ELISA Kit

Brand:  Abnova™ KA0476.1Kit

Additional Details : Weight : 0.48600kg Transport : UN number : 1789 Chem class : 8 Pack group : III


 View more versions of this product

Product Code. 16056115

  • 5943.00 SEK / 1 set


Estimated Shipment
04-02-2021
Add to basket

Includes: Human FBG Microplate: A 96-well polystyrene microplate coated with a polyclonal antibody against human FBG
Sealing Tapes
Human FBG Standard: Human FBG in a buffered protein base (lyophilized)
Biotinylated Human FBG Antibody (50x)
MIX Diluent Concentrate (10x)
Wash Buffer Concentrate (20x)
Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate
Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
Stop Solution: A 0.5N hydrochloric acid

Description

Description

This assay employs a quantitative sandwich enzyme immunoassay technique that measures human FBG in less than 4 hours
Specifications

Specifications

1.25 to 80ng/mL
Human
1ng/mL
Quantification
Antibody coated microwells (96 wells), Human FBG Standard (400ng), Biotinylated FBG Antibody (100x): 1 vial, MIX Diluent Concentrate (10x): 30mL, Wash Buffer Concentrate (20x): 30mL x 2, Streptavidin-Peroxidase Conjugate (SP Conjugate): 80, Chromogen Substrate (8mL), Stop Solution (12mL)
ELISA Kit
RUO
50μL
96 assays
Biotin
Fibrinogen (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human fibrinogen in urine and cell culture supernatant.
Colorimetric
96 wells
Fibrinogen (Human) ELISA Kit
1 Kit
Cell Culture Samples, CSF, Milk, Saliva, Urine
Store the kit at -20°C. The reconstituted standard, SP Conjugate and biotinylated antibody should be stored at -20°C. Others stored at 4°C. Aliquot to avoid repeated freezing and thawing.
50μL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Fibrinogen (Human) ELISA Kit

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P233-Keep container tightly closed.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P404-Store in a closed container.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : proclin 300, sodiumazide, 3,3',5,5'-tetramethylbenzidine, hydrochloric acid, peroxidase stabiliser

SDS
Documents