missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FIGF (Human) ELISA Kit

Brand:  Abnova™ KA2252.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16057165

  • 5943.00 SEK / 1 set


Estimated Shipment
09-02-2021
Add to basket

Includes: 96-Well Microplate or Strips Coated w/Capture Antibody
Biotin-Conjugated Detection Antibody, Lyophilized (2.4µg), Yellow container
Avidin-HRP Conjugate, 10µL, Clear container
Cytokine Protein Standard, Lyophilized (6.5ng), Red container
Ready-to-Use Substrate, 11mL, Brown container
Stop Solution, 11mL, Clear container
Adhesive Plate Sealers, 4 Sheets
Wash Buffer (10X), 50mL, Clear container
Protein Standard Diluent, 15mL, Clear container
Detection Antibody Diluent, 15mL, Clear container
HRP Diluent, 15mL, Clear container

Description

Description

This particular immunoassay utilizes the quantitative technique of a “Sandwich” Enzyme-Linked Immunosorbent Assay (ELISA) where the target protein (antigen) is bound in a “sandwich” format by the primary capture antibodies coated to each well-bottom and the secondary detection antibodies addedsubsequently by the investigator
Specifications

Specifications

Biotin
FIGF (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human FIGF.
Quantification
Antibody coated microwells (96 wells), Biotin-conjugated detection antibody (1 vial), Avidin-HRP conjugate (10μL), Cytokine protein standard (1 vial); Ready-to-use substrate (11mL), Stop solution (11mL), Wash buffer 10X (50mL), Protein standard diluent (15mL), Detection antibody diluent (15mL), HRP diluent (15mL)
ELISA Kit
RUO
100μL
96 Assays
Human
Colorimetric
96 wells
FIGF (Human) ELISA Kit
1 Kit
Cell Lysate, Plasma, Serum
Store the kit at 4°C.
100μL
Videos
SDS
Documents