missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FN1 (Human) ELISA Kit
Brand: Abnova™ KA3039.1Kit
Additional Details : Weight : 0.02000kg
Includes: One 96-well plate pre-coated with anti-Human Fibronectin antibody, 96 (8 x 12) wells
Lyophilized Human Fibronectin standards, 10ng x 2
Sample / Standard diluent buffer, 30mL
Biotin conjugated anti-Human Fibronectin antibody (Concentrated), Dilution: 1:100, 130μL
Antibody diluent buffer, 12mL
Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100, 130μL
ABC diluent buffer, 12mL
TMB substrate, 10mL
Stop solution, 10mL
Wash buffer (25X), 30mL
Description
This kit is based on sandwich enzyme-linked immune-sorbent assay technologySpecifications
156 to 10000pg/mL | |
Human | |
10pg/mL | |
Quantification | |
One 96-well plate pre-coated with anti-Human Fibronectin antibody (96 (8 x 12) wells), Lyophilized Human Fibronectin standards (10ng x 2), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Human Fibronectin antibody (Concentrated), Dilution: 1:100 (130μL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130μL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X): 30mL | |
RUO | |
100μL | |
Store at 4°C for 4 months, or at -20°C for 8 months. | |
100μL |
Biotin | |
FN1 (Human) ELISA Kit is intended for the quantitative measurement of human FN1. | |
Colorimetric | |
96 wells | |
FN1 (Human) ELISA Kit | |
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate | |
<10pg/mL | |
96 Assays |