Learn More
Abnova™ Hepatitis B surface antigen Ab ELISA Kit
Sandwich Enzyme Immunoassay for the measurement of antibodies to Hepatitis B surface antigen
Brand: Abnova™ KA0287.1Kit
Additional Details : Weight : 0.02000kg Transport : UN number : 2796 Chem class : 8 Pack group : II
Specifications
0 to 1000mIU/mL | |
Hepatitis B surface antigen antibody ELISA Kit is a sandwich enzyme immunoassay for the measurement of antibodies to hepatitis B surface antigen. | |
Colorimetric | |
96-well Antibody coated microwell, 6mL HBsAg -Peroxidase solution, 1mL Anti-HBs positive control, 1.5mL HB negative control, 10mL TMB Substrate solution A, 10mL TMB Substrate solution B, 50mL Conc. washing solution D (20X), 10mL 2N Sulfuric acid | |
Hepatitis B surface antigen Ab ELISA kit | |
Plasma, Serum | |
50μL |
HRP | |
10mIU/mL | |
Qualitative | |
Hepatitis B surface antigen Ab ELISA Kit | |
1 Kit | |
Store the kit at 4°C. |