missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IgA (Human) ELISA Kit
Brand: Abnova™ KA2110.1Kit
Additional Details : Weight : 0.02000kg
Includes: Human IgA Microplate: A 96-well polystyrene microplate coated with a polyclonal antibody against human IgA
Sealing Tapes
Human IgA Standard: Human IgA in a buffered protein base, lyophilized, 100ng
Biotinylated Human IgA Antibody (60x): A 60-fold concentrated biotinylated polyclonal antibody against IgA
MIX Diluent Concentrate (10x)
Wash Buffer Concentrate (20x)
Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate
Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
Stop Solution: A 0.5N hydrochloric acid
Description
This assay employs a quantitative sandwich enzyme immunoassay technique that measures human IgA in less than 4 hoursSpecifications
0.781 to 50ng/mL | |
Human | |
0.52ng/mL | |
Quantification | |
Antibody coated microwells (96 wells), Human IgA Standard (200ng), Biotinylated IgA Antibody (50x): 1 vial, MIX Diluent Concentrate (10x): 30mL, Wash Buffer Concentrate (20x): 30mL x 2, Streptavidin-Peroxidase Conjugate (SP Conjugate): 80, Chromogen Substrate (8mL), Stop solution (12mL) | |
ELISA Kit | |
RUO | |
50μL | |
96 Assays |
Biotin | |
IgA (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IgA. | |
Colorimetric | |
96 wells | |
IgA (Human) ELISA Kit | |
1 Kit | |
Cell Culture Sample, CSF, Milk, Plasma, Saliva, Serum, Urine | |
Store SP Conjugate and Biotinylated Antibody and Standard at -20°C. Store all other components at 4°C. | |
50μL |