missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IL18 (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human IL-18
Brand: Abnova™ KA3075.1Kit
Additional Details : Weight : 0.02000kg
Specifications
15.6 to 1000pg/mL | |
Human | |
1pg/mL | |
Quantification | |
IL18 (Human) ELISA Kit | |
RUO | |
100μL | |
96 Assays |
Biotin | |
IL18 (Human) ELISA Kit is intended for the quantitative measurement of human IL18. | |
Colorimetric | |
96-well plate pre-coated with anti-Human IL-18 antibody, Lyophilized IL-18 standards (10ng x 2), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Human IL-18 antibody (Concentrated Dilution: 1:100) (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (ConcentratedDilution: 1:100) (130uL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL) | |
96 Assays | |
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate | |
Store at 4°C for 4 months, or at -20°C for 8 months. | |
100μL |