missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IL18 (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human IL-18
POA
Specifications
Calibration Range | 15.6 to 1000pg/mL |
---|---|
Detection Limit | 1pg/mL |
Detection Method | Colorimetric |
Conjugate | Biotin |
Cross Reactivity | Human |
Specifications
15.6 to 1000pg/mL | |
Colorimetric | |
Human | |
Quantification | |
For research use only (RUO) |
1pg/mL | |
Biotin | |
IL18 (Human) ELISA Kit | |
1 Kit | |
100 μL |