missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IL1A (Human) ELISA Kit

Brand:  Abnova™ KA3055.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16057795

  • 5943.00 SEK / 1 set


Estimated Shipment
10-02-2021
Add to basket

Includes: One 96-well plate pre-coated with anti-Human IL-1α antibody, 96 (8 x 12) wells
Lyophilized IL-1α standards, 10ng x 2
Sample/Standard diluent buffer, 30mL
Biotin conjugated anti-Human IL-1α antibody (Concentrated), Dilution: 1:100, 130μL
Antibody diluent buffer, 12mL
Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100, 130μL
ABC diluent buffer, 12mL
TMB substrate, 10mL
Stop solution, 10mL
Wash buffer (25X), 30mL

Description

Description

This kit is based on sandwich enzyme-linked immune-sorbent assay technology
Specifications

Specifications

4.7 to 300pg/mL
Human
0.5pg/mL
Quantification
One 96-well plate pre-coated with anti-Human IL-1α antibody (96 (8 x 12) wells, Lyophilized IL-1α standards (10ng x 2), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Human IL-1α antibody (Concentrated), Dilution: 1:100 (130μL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130μL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X): 30mL
RUO
100μL
Store at 4°C for 4 months, or at -20°C for 8 months.
100μL
Biotin
IL1A (Human) ELISA Kit is intended for the quantitative measurement of human IL1A.
Colorimetric
96 wells
IL1A (Human) ELISA Kit
Body Fluid, Cell Culture Supernatant, Serum, Tissue Lysate
<0.5pg/mL
96 Assays
Videos
SDS
Documents