Learn More
Abnova™ IL1RN (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human IL-1RN
Brand:  Abnova™ KA1066.1Kit
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16056515
  • 5943.00 SEK / 1 set
Estimated Shipment
12-02-2021
Specifications
31.2 to 2000pg/mL
Human
2pg/mL
Quantification
IL1RN (Human) ELISA Kit
1 Kit
Store the kit at-20°C.
100μL
Biotin
Human IL1RN ELISA kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL1RN.
Colorimetric
96-well Antibody coated microwell, 2 vials x 10ng recombinant Human IL1RN standard (lyophilized), 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Human IL-1RA Antibody, 12mL Antibody Diluent Buffer,, 130μL Avidin-Bi
IL-1RN (Human) ELISA kit
Body Fluid, Cell Culture Supernatant, Serum, Tissue Lysate
96 Assays