missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Lcn2 (Mouse) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Lcn2
POA
Specifications
Detection Method | Colorimetric |
---|---|
Product Type | Lcn2 (Mouse) ELISA Kit |
Includes:
- 96-well plate precoated with anti-mouse NGAL antibody, 96 (8 x 12) wells
- Lyophilized recombinant mouse NGAL standard, 10 ng/tube × 2
- Biotinylated anti- mouse NGAL antibody, dilution 1:100, 130μL
- Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100, 130μL
- Sample diluent buffer, 30mL
- Antibody diluent buffer, 12mL
- ABC diluent buffer, 12mL
- TMB color developing agent, 10mL
- TMB stop solution, 10mL
Specifications
Colorimetric | |
Lcn2 (Mouse) ELISA Kit |