Learn More
Abnova™ Lcn2 (Mouse) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Lcn2
Brand:  Abnova™ KA0559.1Kit
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16056195
  • 5943.00 SEK / 1 set
Estimated Shipment
10-02-2021
Specifications
78 to 5000pg/mL
Mouse
10pg/mL
Quantification
Lcn2 (Mouse) ELISA Kit
1 Kit
Store the kit at-20°C.
100μL
Biotin
Lcn2 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Lcn2.
Colorimetric
96-well plate precoated with Anti-Mouse NGAL Antibody, 2 x 10ng recombinant Mouse NGAL standard (lyophilized), 130μL Biotinylated Anti-Mouse NGAL Antibody, 130μL Avidin-Biotin-Peroxidase Complex (ABC), 30mL Sample Dilue
LCN2 (Mouse) ELISA kit
Cell Culture Supernatant, Plasma (Heparin), Serum, Urine
96 Assays