Learn More
Abnova™ Lcn2 (Mouse) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Lcn2
Brand: Abnova™ KA0559.1Kit
Additional Details : Weight : 0.02000kg
Specifications
78 to 5000pg/mL | |
Mouse | |
10pg/mL | |
Quantification | |
Lcn2 (Mouse) ELISA Kit | |
1 Kit | |
Store the kit at-20°C. | |
100μL |
Biotin | |
Lcn2 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Lcn2. | |
Colorimetric | |
96-well plate precoated with Anti-Mouse NGAL Antibody, 2 x 10ng recombinant Mouse NGAL standard (lyophilized), 130μL Biotinylated Anti-Mouse NGAL Antibody, 130μL Avidin-Biotin-Peroxidase Complex (ABC), 30mL Sample Dilue | |
LCN2 (Mouse) ELISA kit | |
Cell Culture Supernatant, Plasma (Heparin), Serum, Urine | |
96 Assays |