missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ MMP7 (Human) ELISA Kit

Brand:  Abnova™ KA3089.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16057825

  • 5943.00 SEK / 1 set


Estimated Shipment
08-02-2021
Add to basket

Includes: One 96-well plate pre-coated with anti-human MMP-7 antibody, 96 (8 x 12) wells
Standard (13500pg/mL), 0.5mL
Standard diluent buffer, 1.5mL
Wash buffer (30x), Dilution: 1:30, 20mL
Sample diluent buffer, 6mL
HRP conjugated anti-human MMP-7antibody (RTU), 6mL
Stop solution, 6mL
TMB substrate A, 6mL
TMB substrate B, 6mL
Plate sealer, 2 slices
Hermetic bag, 1 bag

Description

Description

This kit is based on standard sandwich enzyme-linked immune-sorbent assay technology
Specifications

Specifications

156 to 10000pg/mL
Human
10pg/mL
Quantification
One 96-well plate pre-coated with anti-human VEGF-D antibody (96 (8x12) wells), Standard (360pg/mL): 0.5mL; Standard diluent buffer (1.5mL), Wash buffer (30x), Dilution: 1:30 (20mL), Sample diluent buffer (6mL), HRP conjugated anti-human VEGF-D antibody (RTU): 6mL, Stop solution (6mL), TMB substrate A (6mL), TMB substrate B (6mL), Plate sealer (2 slices), Hermetic bag (1)
For research use only (RUO)
10μL
96 Assays
HRP
MMP7 (Human) ELISA Kit is intended for the quantitative measurement of human MMP7.
Colorimetric
96 wells
MMP7 (Human) ELISA Kit
Cell Culture Supernatant, Plasma, Serum, Tissue Sample, Urine
Store at 4°C for 6 months.
10μL
Videos
SDS
Documents