missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MMP7 (Human) ELISA Kit
POA
Specifications
Detection Method | Colorimetric |
---|---|
Cross Reactivity | Human |
Product Type | MMP7 (Human) ELISA Kit |
Regulatory Status | For research use only (RUO) |
Sample Volume | 10 μL |
Description
The ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technologySpecifications
Colorimetric | |
MMP7 (Human) ELISA Kit | |
10 μL |
Human | |
For research use only (RUO) |